Learn More
Invitrogen™ RAD26L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589021
Description
Immunogen sequence: TSEERQKMLR DFYASQYPEV KEFFVDSVSQ FNNSSFEKGE QRTRKKSDKR ESLIKPRLSD SETLSFKDST NKISQVCSLK TYKRKSVKFQ NHISYREEVF FNDAETKKSP VSSTQEIDSG KNSQASEDTV TSRSLNSESE TRERRLENTM KDQQDLTRTG ISRKEPLLKL ENKKIENPVL ENTSVISLLG DTSILDDLFK SHGNSPTQLP KKVLSGPMEK AKQRPKDFWD ILNEQNDESL SKLTDLAVIE TLCEKAPLAA PFKRREEPAT SLWKSNEKFL WKKFSPSDTD ENATNTQSTT; Positive Samples: U-87MG, HeLa, LO2, MCF-7, Mouse testis, Mouse kidney; Cellular Location: Cytoplasm, Mitochondrion, Nucleus, centrosome, cytoskeleton, microtubule organizing center.
This gene encodes a member of the Snf2 family of helicase-like proteins. The encoded protein may play a role in DNA repair and mitochondrial function. Mutations in this gene have been associated with bone marrow failure syndrome 2. Alternatively spliced transcript variants that encode different protein isoforms have been described.
Specifications
| RAD26L | |
| Polyclonal | |
| Unconjugated | |
| Ercc6l2 | |
| 0610007P08Rik; 1700019D06Rik; 9330134C04Rik; BMFS2; C8H9orf102; C9orf102; DNA excision repair protein ERCC-6-like 2; DNA repair and recombination protein RAD26-like; ERCC excision repair 6 like 2; Ercc6l2; excision repair cross-complementation group 6 like 2; excision repair cross-complementation group 6-like 2; excision repair cross-complementing rodent repair deficiency, complementation group 6 like 2; excision repair cross-complementing rodent repair deficiency, complementation group 6-like 2; LOW QUALITY PROTEIN: DNA excision repair protein ERCC-6-like 2; putative DNA repair and recombination protein RAD26-like; putative DNA repair and recombination protein RAD26-like-like protein; putative repair and recombination helicase RAD26L; RAD26L; RGD1561537; Sr278; stretch responsive protein 278 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 375748, 76251 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.59 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5T890, Q9JIM3 | |
| Ercc6l2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1262-1561 of human ERCC6L2 (NP_0645922). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.