missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD23B Recombinant Rabbit Monoclonal Antibody (SAIC-28A-29)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547909
This item is not returnable.
View return policy
Description
Specificity: Recognises human RAD23B.
The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in the nucleotide excision repair (NER). This protein was found to be a component of the protein complex that specifically complements the NER defect of xeroderma pigmentosum group C (XP-c) cell extracts in vitro. This protein was also shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, and thus this protein may be involved in the ubiquitin mediated proteolytic pathway in cells.
Specifications
| RAD23B | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P54727 | |
| Rad23b | |
| Peptide ILNDDTALK, derived from the UV excision repair protein RAD23 homolog B, conjugated to KLH. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| Flow Cytometry, Western Blot, Immunocytochemistry, Mass Spectrometry | |
| SAIC-28A-29 | |
| Unconjugated | |
| Rad23b | |
| 0610007D13Rik; AV001138; HHR23B; HR23B; Mhr23b; P58; RAD23 homolog B (S. cerevisiae); RAD23 homolog B, nucleotide excision repair protein; RAD23, yeast homolog of, B; RAD23B; RAD23b homolog; RAD23b homolog (S. cerevisiae); UV excision repair protein RAD23 homolog B; XP-C repair complementing complex 58 kDa; XP-C repair complementing protein; XP-C repair-complementing complex 58 kDa protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5887 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction