missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD23A Monoclonal Antibody (1E4D6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531886
This item is not returnable.
View return policy
Description
MA5-31886 has been tested in indirect ELISA.
The product of this gene is a small iron-sulfur protein that transfers electrons from NADPH through ferredoxin reductase to a terminal cytochrome P450. This particular oxidation/reduction system is found in steroidogenic tissues, and is involved with the synthesis of bile acid and vitamin D. In addition to the expressed gene at this chromosomal locus (11q22), there are pseudogenes located on chromosomes 20 and 21. This gene product has been identified in a number of different tissues but all forms have been shown to be identical and are not tissue specific.
Specifications
| RAD23A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P54725 | |
| RAD23A | |
| Purified recombinant fragment of human RAD23A (AA: 1-363) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Western Blot | |
| 1E4D6 | |
| Unconjugated | |
| RAD23A | |
| 2310040P19Rik; AL024030; HHR23A; HR23A; MGC111083; Mhr23a; rad23; RAD23 homolog A (S. cerevisiae); RAD23 homolog A, nucleotide excision repair protein; RAD23, yeast homolog, A; RAD23A; RAD23a homolog (S. cerevisiae); UV excision repair protein RAD23 homolog A | |
| Mouse | |
| Protein G | |
| RUO | |
| 5886 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction