missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad21 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-203H
This item is not returnable.
View return policy
Description
Rad21 Polyclonal antibody specifically detects Rad21 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Rad21 | |
| Polyclonal | |
| Western Blot | |
| double-strand-break repair protein rad21 homolog, hHR21FLJ25655, HR21, KIAA0078RAD21 (S. pombe) homolog, MCD1, Nuclear matrix protein 1, NXP-1, NXP1HRAD21, protein involved in DNA double-strand break repair, RAD21 homolog (S. pombe), SCC1 homolog, SCC1FLJ40596 | |
| The immunogen recognized by this antibody maps to a region between residue 575 and the C-terminus (residue 631) human Rad21 homolog using the numbering given in entry NP_006256.1 (GeneID 5885). | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair | |
| 5885 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction