missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad21 Antibody (CM110-2C10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-386CL1
This item is not returnable.
View return policy
Description
Rad21 Monoclonal antibody specifically detects Rad21 in Human,Mouse samples. It is validated for Western Blot,Simple Western
Specifications
| Rad21 | |
| Monoclonal | |
| CoraFluor 1 | |
| double-strand-break repair protein rad21 homolog, hHR21FLJ25655, HR21, KIAA0078RAD21 (S. pombe) homolog, MCD1, Nuclear matrix protein 1, NXP-1, NXP1HRAD21, protein involved in DNA double-strand break repair, RAD21 homolog (S. pombe), SCC1 homolog, SCC1FLJ40596 | |
| An N-terminal fusion protein of the C-terminal fragment of Scc1 subunit human cohesin (amino acids 544-631 of hRad21). | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair | |
| 5885 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Western Blot, Simple Western | |
| CM110-2C10 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction