Learn More
Invitrogen™ RAD18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120911
Description
Positive test controls include: MCF7. The target is usually found in the following locations: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center. Immunogen sequence: INKHLDSCLS REEKKESLRS SVHKRKPLPK TVYNLLSDRD LKKKLKEHGL SIQGNKQQLI KRHQEFVHMY NAQCDALHPK SAAEIVREIE NIEKTRMRLE ASKLNESVMV FTKDQTEKEI DEIHSKYRKK HKSEFQLLVD QARKGYKKIA GMSQKTVTIT KEDESTEKLS SVCMGQEDNM TSVTNHFSQS KLDSPEELEP DREEDSSSCI DIQEVLSSSE SDSCNSSSSD IIRDLLEEEE AWEASHKNDL QDTEISPRQN RRTRAAESAE IEPRNKRNRN.
Human Rad18 (hRad18) is homologue of the S. pombe Rad18 (3,4). In yeast, Rad6 and Rad18 play a crucial role in post-replication repair functions in gap-filling of a daughter strand on replication of damaged DNA (2). Furthermore, Rad6-Rad18 interaction is thought to be critical at an early step during lesion bypass in yeast. In vivo, Rad18 protein forms a tight complex with Rad6 through a conserved ring-finger motif (3,4). It has been suggested that Rad18-Rad6 complex binds to the damaged sites by the single-stranded DNA binding property of Rad 18. This is followed by degradation of the stalled components of DNA replication by ubiquitin-conjugating enzyme activity of Rad6 (1). The human Rad18 maps on chromosome 3p24-25, the region which is often deleted in lung, breast, ovary, and testis cancers. The human RAD18 gene codes for a protein of 495 amino acids with a predicted molecular weight of 56 kDa. The human Rad18 is ubiquitously expressed in various human tissues.
Specifications
| RAD18 | |
| Polyclonal | |
| Unconjugated | |
| Rad18 | |
| 2810024C04Rik; E3 ubiquitin-protein ligase RAD18; hHR18; hRAD18; mRAD18Sc; postreplication repair protein hRAD18p; postreplication repair protein RAD18; post-replication repair protein RAD18SC; Rad18; RAD18 E3 ubiquitin protein ligase; RAD18 homolog; RAD18 homolog (S. cerevisiae); RAD18, E3 ubiquitin protein ligase; RAD18, S. cerevisiae, homolog; Rad18sc; RING finger protein 73; RING-type E3 ubiquitin transferase RAD18; RNF73 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56852 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NS91 | |
| Rad18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 216-495 of human Rad18 (NP_064550.3). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.