missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38140-100ul
This item is not returnable.
View return policy
Description
RAD18 Polyclonal antibody specifically detects RAD18 in Human samples. It is validated for ELISA,Western Blot
Specifications
| RAD18 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 6.3.2.-, hHR18, hRAD18, postreplication repair protein hRAD18p, Postreplication repair protein RAD18, RAD18 homolog (S. cerevisiae), RAD18, S. cerevisiae, homolog, RING finger protein 73, RNF73E3 ubiquitin-protein ligase RAD18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 216-495 of human RAD18 (NP_064550.3).,, Sequence:, INKHLDSCLSREEKKESLRSSVHKRKPLPKTVYNLLSDRDLKKKLKEHGLSIQGNKQQLIKRHQEFVHMYNAQCDALHPKSAAEIVREIENIEKTRMRLEASKLNESVMVFTKDQTEKEIDEIHSKYRKKHKSEFQLLVDQARKGYKKIAGMSQKTVTITKEDESTEKLSSVCMGQEDNMTSVTNHFSQSKLDSPEELEPDREEDSSSCIDIQEVLSSSESDSCNSSSSDIIRDLLEEEEAWEASHKNDLQDTEISPRQNRRTRAAESAEIEPRNKRNRN | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers, Zinc Finger | |
| 56852 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction