missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD1 Monoclonal Antibody (4126)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA123350
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2.
Rad1 is a component of a heterotrimeric PCNA-like complex that contains Rad9 and Hus1 proteins. This complex is involved in the 9-1-1 cell cycle checkpoing response complex that plays a major role in DNA repair, by associating with Rad17 and several components of the PCNA-loading heteropentamer replication factor C. Mutants has been shown to partially restore radiation resistance and G2-checkpoint activity. It has also been shown to possess exonuclease activity.
Specifications
| RAD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 20% glycerol and no preservative; pH 7 | |
| O60671 | |
| RAD1 | |
| Amino terminal fragment of human Rad1 purified from E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 4126 | |
| Unconjugated | |
| RAD1 | |
| cell cycle checkpoint protein Hrad1; cell cycle checkpoint protein RAD1; checkpoint control protein HRAD1; DNA repair exonuclease rad1 homolog; DNA repair exonuclease REC1; exonuclease homolog RAD1; hRAD1; mRAD1; RAD1; RAD1 checkpoint clamp component; RAD1 checkpoint DNA exonuclease; RAD1 homolog; RAD1 homolog (S. pombe); rad1-like DNA damage checkpoint protein; REC1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5810 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction