missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97717JF646
This item is not returnable.
View return policy
Description
Rad1 Polyclonal specifically detects Rad1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Rad1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| cell cycle checkpoint protein Hrad1, checkpoint control protein HRAD1, DNA repair exonuclease rad1 homolog, DNA repair exonuclease REC1, EC 3.1.11.2, exonuclease homolog RAD1, HRAD1, RAD1 (S. pombe) homolog, RAD1 homolog (S. pombe), Rad1-like DNA damage checkpoint protein, REC1cell cycle checkpoint protein RAD1 | |
| Produced in rabbits immunized with E. coli-derived Human Rad1 fragment. | |
| 0.1 mL | |
| Cancer, DNA Repair, Signal Transduction | |
| 5810 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction