missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad1 Antibody (1G2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00005810-M01J-0.1mg
This item is not returnable.
View return policy
Description
Rad1 Monoclonal antibody specifically detects Rad1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA, Sandwich ELISA Capture, KnockDown
Specifications
| Rad1 | |
| Monoclonal | |
| Unconjugated | |
| 1X PBS pH 7.4 | |
| cell cycle checkpoint protein Hrad1, checkpoint control protein HRAD1, DNA repair exonuclease rad1 homolog, DNA repair exonuclease REC1, EC 3.1.11.2, exonuclease homolog RAD1, HRAD1, RAD1 (S. pombe) homolog, RAD1 homolog (S. pombe), Rad1-like DNA damage checkpoint protein, REC1cell cycle checkpoint protein RAD1 | |
| RAD1 (AAH06837, 1 a.a. ∽ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPLLTQQIQDEDDQYSLVASLDNVRNLSTILKAIHFREHATCFATKNGIKVTVENAKCVQANAFIQAGIFQEFKVQEESVTFRINLTVLL | |
| 0.1 mg | |
| Cancer, DNA Repair, Signal Transduction | |
| 5810 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG3 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA, Sandwich ELISA Capture, KnockDown | |
| 1G2 | |
| Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA, Sandwich ELISA Capture, Knockdown Validated | |
| AAH06837 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction