Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Brand:  Thermo Scientific™ 1600006
Code : A0
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10458434
  • £123.91 / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket
Description
Description
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Specifications
Specifications
Rack Insert; Includes top and bottom panel with no holes
Bath Types A5B, A10B, A24B, S49, S19T, S14P, S21P
Rack
Stainless Steel