Learn More
Hamilton™ Non-Filtered Racked Pipette Tips
Delivering outstanding accuracy and precision, guaranteed.
Brand:  Hamilton™ 68008-12
Code : NEW
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15305094
  • £68.65 / Pack of 10
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
  • Pre-sterilized or non-sterile
  • 96 tips to a rack
Specifications
Specifications
200 μL
Non-Filtered
Sterile
Virgin Polypropylene