missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RACK1 Recombinant Rabbit Monoclonal Antibody (JG40-26)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534809
This item is not returnable.
View return policy
Description
Positive Control: SH-SY-5Y, rat small intestine tissue lysate, rat cerebellum tissue, rat liver tissue, human colon tissue, mouse skin tissue.
RACK1 (receptor for activated C kinase 1) was identified through its binding to various PKC isoforms. Its main function is to recruit PKC and various other proteins to specific locations to form multiprotein complexes, mediating various signal pathways. RACK1 is required for VANGL2 membrane localization and for PTK2/FAK1 phosphorylation and dephosphorylation.
Specifications
| RACK1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
| P63244, P63245, P68040 | |
| RACK1 | |
| Recombinant protein within Human RACK1 aa 190-317. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JG40-26 | |
| Unconjugated | |
| RACK1 | |
| 12-3; activated protein kinase C receptor; AL033335; Cell proliferation-inducing gene 21 protein; CG7111; CG7111-PA; CG7111-PB; CG7111-PC; CG7111-PD; Dmel\CG7111; Dmel_CG7111; dRack1; GB-like; gnb2l1; Gnb2-rs1; guanine nucleotide binding protein (G protein) beta polypeptide 2-like 1; guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1; guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1 sequence 1; guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1; guanine nucleotide binding protein beta polypeptide 2-like 1; guanine nucleotide binding protein related; guanine nucleotide binding protein, beta 2, related sequence 1; guanine nucleotide binding protein, beta polypeptide 2-like 1; guanine nucleotide binding protein, beta-2, related sequence 1; guanine nucleotide-binding protein beta-subunit like protein; guanine nucleotide-binding protein subunit beta-2-like 1; Guanine nucleotide-binding protein subunit beta-2-like 1, N-terminally processed; Guanine nucleotide-binding protein subunit beta-like protein; Guanine nucleotide-binding protein subunit beta-like protein 12.3; H12.3; HLC7; HLC-7; Human lung cancer oncogene 7 protein; i173; lung cancer oncogene 7; OTTHUMP00000223870; OTTHUMP00000223891; OTTHUMP00000223893; OTTHUMP00000223900; OTTHUMP00000223901; OTTHUMP00000223902; OTTHUMP00000223930; OTTHUMP00000223931; p205; PIG21; proliferation-inducing gene 21; protein homologous to chicken B complex protein, guanine nucleotide binding; protein kinase C receptor; RACK; Rack1; rack-1; Rack1-PA; Rack1-PB; Rack1-PC; Rack1-PD; RE74715p; receptor for activated C kinase; receptor for activated C kinase 1; receptor for activated protein kinase C; receptor for activated protein kinase C1; receptor of activated PKC; receptor of activated protein C kinase 1; Receptor of activated protein C kinase 1, N-terminally processed; Receptor of activated protein kinase C; receptor of activated protein kinase C 1; Receptor of activated protein kinase C homolog; similar to mammalian RACK1; Small ribosomal subunit protein RACK1; wu:fb80d08; wu:fk65d12 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10399, 14694, 83427 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction