missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RACK1/GNB2L1 Antibody (OTI2D8), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73781AF532
This item is not returnable.
View return policy
Description
RACK1/GNB2L1 Monoclonal specifically detects RACK1/GNB2L1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| RACK1/GNB2L1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| GNB2L1 | |
| Full length recombinant protein of human RACK1/GNB2L1 (NP_006089) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2D8 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| guanine nucleotide binding, guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1, Receptor for activated C kinase, Receptor for Activated C Kinase 1 | |
| Mouse | |
| 0.1 mL | |
| 10399 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction