Learn More
Edmund Bühler GmbH™ Combifix VKS B Rack System
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
SGM-590 Series Edmund Bühler
missing translation for 'unitSize'
Each
Description
- For use with Combifix VKS B
- Includes:
- 4 clamping strips h
- 4 clampings strips v
- For fastening horizontal or high vessels between the clamping strips
- Distance between the strips h and v approximately 60mm
- To be mounted to a shaking plate
Specifications
Specifications
| Product Type | Rack System Combifix B |
| For Use With (Equipment) | SGM-590 Series Edmund Bühler |
| Includes | Basic Rack, Clamping Strips 4 x h and 4 x v |