Learn More
Memmert™ Rack for shaking device for 475 test tubes
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB35
WTB50
Unit Size:
Each
Description
- Rack for shaking device for test tubes
- Accessory
Specifications
Specifications
| For Use With (Equipment) | WTB35 |
| Type | Rack for shaking device for 475 test tubes 14.5 mm |
| Diameter (Metric) | 14.5 mm |