Learn More
Tenak™ Eco Alurack for 20 mm Microtiterplates
Product Code. 15250994 Shop All Tenak Products
Click to view available options
Holds:
Microtiterplates (114)
Unit Size:
Each
Product Code. 15250994
Brand: Tenak™ TE27713
Only null left
Add to basket
Add to basket
Freezer rack designed to hold microtiterplates.
  • Rack
  • For use with freezers
Specifications
For Use With (Equipment) Freezers
Holds Microtiterplates (114)
Array 19 x 6