missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Cardboard Rack for Upright Freezers, 50 mm Box Height
Due to the design and choice of material, these unique racks are extremely solid and long lasting.
Brand: Tenak™ TE26132
Additional Details : Weight : 1.10000kg
Description
- Gives the end user a second to none opportunity for reducing costs
- Ideal for long term storage or for applications where price is of high priority
Specifications
50 mm (Box) | |
Upright Freezers | |
4 x 4 |
562 x 139 x 225 mm | |
Boxes (16) | |
Cardboard |