missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Freezer Rack for 42 Matrix Plates

Product Code. 12406980 Shop All Tenak Products
Click to view available options
Holds:
Matrix Plates (42)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12406980

Brand: Tenak™ TE27712

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Freezer rack designed to hold matrix plates.

  • Rack
  • For use with freezers

Specifications

Array 6 x 7
Holds Matrix Plates (42)
For Use With (Equipment) Freezers
Height (Metric) 42 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.