Learn More
Tenak™ Stainless Steel Freezer Rack for 24 Microtiterplates
Shop All Tenak Products
Click to view available options
Holds:
Microtiterplates (24)
Unit Size:
Each
Description
- Rack
- For use with freezers
Specifications
Specifications
| Array | 4 x 6 |
| Holds | Microtiterplates (24) |
| Material | Stainless Steel |
| For Use With (Equipment) | Freezers |
| Height (Metric) | 51 mm |