Learn More
Greiner Bio-One Rack with Cryo.s Biobanking Tubes and Capping Tool
Description
Rack:
- Stackable, with footprint
- Rack bottom with scanning windows for Datamatrix, rotation stoppers, snap-in lid
Tubes:
- Fitted with screw caps (made of silicone gasket and inte
Compliance
Corresponds to American National Standards Institute (ANSI-Standard)
Specifications
Specifications
| Color | Black |
| Capacity (Metric) | 300 μL |
| Material | Polycarbonate |
| Packaging | 5 Racks/Bag |
| RNase Free | Yes |
| Sterility | Sterile |
| Graduated | Yes |
| Closure Type | Silicone Gasket, Internal Thread |