Learn More
Greiner Bio-One Rack with Cryo.s Biobanking Tubes and Capping Tool
GREENER_CHOICE
Product Code. 15756242
Click to view available options
Capacity (Metric):
600 μL
Packaging:
2 Racks/Bag
Unit Size:
Pack of 960
Product Code. 15756242
Brand: Greiner Bio-One 977561
Optimized for automated laboratory equipment such as decappers, liquid handling systems and automated storage systems.
Rack:
  • Stackable, with footprint
  • Rack bottom with scanning windows for Datamatrix, rotation stoppers, snap-in lid
Tubes:
  • Fitted with screw caps (made of silicone gasket and inte
Compliance
Corresponds to American National Standards Institute (ANSI-Standard)
TRUSTED_SUSTAINABILITY
Specifications
Product Type CryoTube with Rack
Capacity (Metric) 600 μL
Material Polycarbonate
No. per Pack 96
Packaging 2 Racks/Bag
RNase Free Yes
Sterility Sterile
Graduated Yes
Closure Type Silicone Gasket, Internal Thread