Learn More
RACGAP1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30042-1-AP-20UL
Description
The RACGAP1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RACGAP1. This antibody recognizes human antigen. The RACGAP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| RACGAP1 | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| HsCYK 4, ID GAP, KIAA1478, Male germ cell RacGap, MgcRacGAP, RACGAP1 | |
| 20 μL | |
| 29127 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |