missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RACGAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583692
This item is not returnable.
View return policy
Description
Immunogen sequence: SALAFLNRGQ PSSSNAGNKR LSTIDESGSI LSDISFDKTD ESLDWDSSLV KTFKLKKREK RRSTSRQFVD GPPGPVKKTR SIGSAVDQGN ESIVAKTTVT VPNDGGPIEA VSTIET Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 90%.
RAC, a small GTPase in the ras family, regulates the formation of membrane ruffles, lamellipodia and filopodia. It is involved in cytoskeletal actin organization and transformation. The p21 activated kinases (PAKs) are direct targets of active Rac and Cdc42 which can induce the assembly of polarized cytoskeletal structures when expressed in fibroblasts. RAC1 and RAC2 are also essential components of NADPH oxidase, the enzyme responsible for generating free radicals.
Specifications
| RACGAP1 | |
| Polyclonal | |
| Unconjugated | |
| RACGAP1 | |
| AI227039; AI327394; AKT; Band25; CYK4; GAP; gtl11; GTPase; HsCYK-4; ID; ID-GAP; KIAA1478; Male germ cell RacGap; MGC99656; MgcRacGAP; mKIAA1478; PKB; PKB-ALPHA; PRKBA; protein CYK4 homolg; Protein CYK4 homolog; RAC; Rac GTPase activating protein 1; Rac GTPase-activating protein 1; RAC-ALPHA; Racgap1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29127 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H0H5 | |
| RACGAP1 | |
| Recombinant protein corresponding to Human RACGAP1. Recombinant protein control fragment (Product #RP-97840). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction