missing translation for 'onlineSavingsMsg'
Learn More
Learn More
racemic Juvenile hormone III acid, TRC
Click to view available options
Quantity:
2.5 mg
10 mg
Unit Size:
10mg
2.5mg
Specifications
Specifications
| Chemical Name or Material | racemic Juvenile hormone III acid |
| CAS | 10147-50-9 |
| Molecular Formula | C15H24O3 |
| Quantity | 10 mg |
| Synonym | (E,E)-10,11-epoxy-3,7,11-trimethyl-2,6-Dodecadienoic acid (8CI), (2E,6E)-9-(3,3-dimethyloxiranyl)-3,7-dimethyl-2,6-Nonadienoic acid (9CI), (E,E)-9-(3,3-dimethyloxiranyl)-3,7-dimethyl-2,6-Nonadienoic acid, (2E,6E)-9-(3,3-Dimethyl-2-oxiranyl)-3,7-dimethyl-2,6-nonadienoic acid, (E,E)-3,7,11-Trimethyl-10,11-epoxy-2,6-dodecadienoic acid, JH III acid |
| InChI Formula | InChI=1S/C15H24O3/c1-11(8-9-13-15(3,4)18-13)6-5-7-12(2)10-14(16)17/h6,10,13H,5,7-9H2,1-4H3,(H,16,17)/b11-6+,12-10+ |
| SMILES | C\C(=C/CC\C(=C\C(=O)O)\C)\CCC1OC1(C)C |
| IUPAC Name | (2E,6E)-9-(3,3-dimethyloxiran-2-yl)-3,7-dimethylnona-2,6-dienoic acid |
| Molecular Weight (g/mol) | 252.35 |
| Formula Weight | 252.1725 g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction