missing translation for 'onlineSavingsMsg'
Learn More

Racecadotril, MedChemExpress

Product Code. 16431256
Encompass_Preferred
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
This item is not returnable. View return policy

Product Code. 16431256

Brand: Medchem Express HY17399

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Racecadotril (Acetorphan) is a neutral endopeptidase (NEP) inhibitor. Racecadotril and its active metabolite Thiorphan inhibits purified NEP activity from mouse brain with Kis of 4500 and 6.1 nM, , respectively. Antidiarrheal agent.

Racecadotril (Acetorphan) is a neutral endopeptidase (NEP) inhibitor. Racecadotril and its active metabolite Thiorphan inhibits purified NEP activity from mouse brain with Kis of 4500 and 6.1 nM, , respectively. Antidiarrheal agent.

Specifications

Chemical Name or Material Racecadotril
Physical Form Solid
CAS 81110-73-8
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C21H23NO4S
For Use With (Application) Neuroscience-Neuromodulation
Synonym Acetorphan
Solubility Information DMSO : ≥ 100 mg/mL (259.42 mM)
SMILES O=C(OCC1=CC=CC=C1)CNC(C(CSC(C)=O)CC2=CC=CC=C2)=O
Molecular Weight (g/mol) 385.48
Formula Weight 385.48
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.