missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAC2 Mouse anti-Human, Mouse, Clone: 4C2H6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60077-1-IG-150UL
This item is not returnable.
View return policy
Description
RAC, a small GTPase in the ras family, regulates the formation of membrane ruffles, lamellipodia and filopodia. It is involved in cytoskeletal actin organization and transformation. The p21 activated kinases (PAKs) are direct targets of active Rac and Cdc42 which can induce the assembly of polarized cytoskeletal structures when expressed in fibroblasts. RAC1 and RAC2 are also essential components of NADPH oxidase, the enzyme responsible for generating free radicals.Specifications
| RAC2 | |
| Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P15153, Q05144 | |
| RAC2 | |
| RAC2 Fusion Protein Ag1202 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 4C2H6 | |
| Unconjugated | |
| RAC2 | |
| EN 7, Gx, HSPC022, p21 Rac2, RAC2, Small G protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 19354, 5880 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction