Learn More
Rac1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-24072-20UL
Description
The Rac1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Rac1. This antibody recognizes human, mouse, rat antigen. The Rac1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Rac1 | |
| 1000 μg/mL | |
| MIG5, p21 Rac1, RAC1, RAC1b, Ras like protein TC25, TC 25, TC25 | |
| 20 μL | |
| 5879 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |