missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544839
This item is not returnable.
View return policy
Description
Peptide sequence: LAMAKEIGAV KYLECSALTQ RGLKTVFDEA IRAVLCPPPV KKRKRKCLLL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 100%.
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| RAC1 | |
| Polyclonal | |
| Unconjugated | |
| Rac1 | |
| AL023026; cell migration-inducing gene 5 protein; D5Ertd559e; GTPase; MGC111543; MIG5; p21-Rac1; Rac 1b; Rac family small GTPase 1; RAC1; Rac-1; Rac2; rac2 GTP-binding protein; ras-like protein TC25; RAS-related C3 botulinum substrate 1; ras-related C3 botulinum toxin substrate 1; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1); rho family, small GTP binding protein Rac1; TC25; TC-25 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5879 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P63000 | |
| Rac1 | |
| Synthetic peptide directed towards the middle region of human RAC1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction