missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rac1 [p Ser71] Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38637-20ul
This item is not returnable.
View return policy
Description
Rac1 Polyclonal antibody specifically detects Rac1 in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Rac1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| Cell migration-inducing gene 5 protein, MGC111543, p21-Rac1migration-inducing gene 5, Ras-like protein TC25, ras-related C3 botulinum toxin substrate 1, ras-related C3 botulinum toxin substrate 1 (rho family, small GTP bindingprotein Rac1), rho family, small GTP binding protein Rac1, TC25, TC-25 | |
| A synthetic phosphorylated peptide around S71 of human Rac1 (NP_008839.2).,, Sequence:, PLSYP | |
| 20 μL | |
| Breast Cancer, Cancer, mTOR Pathway, Phospho Specific | |
| 5879 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction