missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rac1 Mouse anti-Human, Clone: CPTC-RAC1-1, Novus Biologicals™
Mouse Monoclonal Antibody
£396.00
Specifications
| Antigen | Rac1 |
|---|---|
| Clone | CPTC-RAC1-1 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin, Protein Array |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
Rac1 Monoclonal specifically detects Rac1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Rac1 | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, mTOR Pathway, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 5879 | |
| IgG1 κ | |
| Protein A or G purified |
| CPTC-RAC1-1 | |
| Western Blot 1 to 2 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell migration-inducing gene 5 protein, MGC111543, p21-Rac1migration-inducing gene 5, Ras-like protein TC25, ras-related C3 botulinum toxin substrate 1, ras-related C3 botulinum toxin substrate 1 (rho family, small GTP bindingprotein Rac1), rho family, small GTP binding protein Rac1, TC25, TC-25 | |
| Recombinant human full-length Rac1protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title