missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Rac1 Polyclonal specifically detects Rac1 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Rac1 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | Cell migration-inducing gene 5 protein, MGC111543, p21-Rac1migration-inducing gene 5, Ras-like protein TC25, ras-related C3 botulinum toxin substrate 1, ras-related C3 botulinum toxin substrate 1 (rho family, small GTP bindingprotein Rac1), rho family, small GTP binding protein Rac1, TC25, TC-25 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Rac1 (NP_008839.2; Met1-Cys189). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?