missing translation for 'onlineSavingsMsg'
Learn More

Rac1,2,3 Antibody (AT2G10), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18422116 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422116 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422116 Supplier Novus Biologicals Supplier No. NBP288839AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rac1,2,3 Monoclonal antibody specifically detects Rac1,2,3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rac1,2,3
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone AT2G10
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias AI323801;AI452260;AL023026;D5Ertd559e;EN-7;Gx;HSPC022;MIG5;p21-Rac1;p21-Rac2;RAC1;Rac-1;Rac1B;RAC2;RAC3;TC25;TC-25
Host Species Mouse
Immunogen Recombinant human Rac1,2,3 (1-189aa) protein purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cytoskeleton Markers, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.