missing translation for 'onlineSavingsMsg'
Learn More

rac,trans-Cevimeline Hydrochloride, TRC

Product Code. 30359662
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359662 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359662 Supplier Toronto Research Chemicals Supplier No. TRCC28352510MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac,trans-Cevimeline Hydrochloride
CAS 107220-29-1
Molecular Formula C10H18ClNOS
Quantity 10 mg
Synonym trans-2'-Methyl-spiro[1-azabicyclo[2.2.2]octane-3,5'-[1,3]oxathiolane] Hydrochloride, trans-2-Methylspiro[1-azabicyclo[2.2.2]octane-3,5-[1,3]oxathiolane] Hydrochloride
InChI Formula InChI=1S/C10H17NOS.ClH/c1-8-12-10(7-13-8)6-11-4-2-9(10)3-5-11;/h8-9H,2-7H2,1H3;1H/t8-,10+;/m0./s1
SMILES Cl.C[C@H]1O[C@@]2(CS1)CN3CCC2CC3
IUPAC Name (2S,5R)-2-methylspiro[1,3-oxathiolane-5,3'-1-azabicyclo[2.2.2]octane];hydrochloride
Molecular Weight (g/mol) 235.77
Formula Weight 235.0798
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.