missing translation for 'onlineSavingsMsg'
Learn More

rac-trans-7-Hydroxy Pramipexole, TRC

Product Code. 30401610
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401610 10 mg 10mg
30404280 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401610 Supplier Toronto Research Chemicals Supplier No. TRCH95214510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246818-51-8
Molecular Formula C10 H17 N3 O S
Molecular Weight (g/mol) 227.33
Synonym 7-Benzothiazolol, 2-amino-4,5,6,7-tetrahydro-6-(propylamino)-, (6R,7S)-rel-, rel-(6R,7S)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)-7-benzothiazolol, (6SR,7RS)-2-Amino-6-(propylamino)-4,5,6,7-tetrahydrobenzothiazol-7-ol, rac-trans-7-Hydroxypramipexole, (±)-trans-7-Hydroxypramipexole
IUPAC Name (6S,7R)-2-amino-6-(propylamino)-4,5,6,7-tetrahydro-1,3-benzothiazol-7-ol
SMILES CCCN[C@H]1CCc2nc(N)sc2[C@@H]1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H17N3OS/c1-2-5-12-6-3-4-7-9(8(6)14)15-10(11)13-7/h6,8,12,14H,2-5H2,1H3,(H2,11,13)/t6-,8+/m0/s1
Formula Weight 227.1092
Chemical Name or Material rac-trans-7-hydroxy Pramipexole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.