missing translation for 'onlineSavingsMsg'
Learn More

rac trans-4-Hydroxy Glyburide, TRC

Product Code. 30391551
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391551 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391551 Supplier Toronto Research Chemicals Supplier No. TRCH94262525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac trans-4-Hydroxy Glyburide
CAS 23155-00-2
Molecular Formula C23 H28 Cl N3 O6 S
Quantity 25 mg
Synonym Benzamide, 5-chloro-N-[2-[4-[[[[(4-hydroxycyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-2-methoxy-, trans-, Urea, 1-[[p-[2-(5-chloro-o-anisamido)ethyl]phenyl]sulfonyl]-3-(4-hydroxycyclohexyl)-, trans- (8CI), 5-Chloro-N-[2-[4-[[[[(trans-4-hydroxycyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-2-methoxybenzamide, 4-trans-Hydroxycyclohexyl glyburide, 4-trans-Hydroxyglibenclamide, N-[4-[β-(2-Methoxy-5-chlorobenzamido)ethyl]benzenesulfonyl]-N'-(trans-4-hydroxycyclohexyl)urea, trans-4-Hydroxyglyburide, trans-N-4-[2-(5-Chloro-2-methoxybenzamido)ethyl]phenylsulfonyl-N'-(4-hydroxycyclohexyl)urea, Benzamide, 5-chloro-N-[2-[4-[[[[(trans-4-hydroxycyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-2-methoxy-, trans-4-Hydroxyglibenclamide
InChI Formula InChI=1S/C23H28ClN3O6S/c1-33-21-11-4-16(24)14-20(21)22(29)25-13-12-15-2-9-19(10-3-15)34(31,32)27-23(30)26-17-5-7-18(28)8-6-17/h2-4,9-11,14,17-18,28H,5-8,12-13H2,1H3,(H,25,29)(H2,26,27,30)/t17-,18-
SMILES COc1ccc(Cl)cc1C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)N[C@@H]3CC[C@@H](O)CC3
IUPAC Name 5-chloro-N-[2-[4-[(4-hydroxycyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]-2-methoxybenzamide
Molecular Weight (g/mol) 510
Formula Weight 509.1387
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.