missing translation for 'onlineSavingsMsg'
Learn More

rac-trans 3’-Hydroxymethylnicotine Hemisuccinate, TRC

Product Code. 30398800
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398800 10 mg -
30403265 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398800 Supplier Toronto Research Chemicals Supplier No. TRCH94819010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1207282-59-4
Molecular Formula C15 H20 N2 O4
Molecular Weight (g/mol) 292.33
IUPAC Name 4-[[(2S,3S)-1-methyl-2-pyridin-3-ylpyrrolidin-3-yl]methoxy]-4-oxobutanoic acid
SMILES CN1CC[C@H](COC(=O)CCC(=O)O)[C@H]1c2cccnc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H20N2O4/c1-17-8-6-12(10-21-14(20)5-4-13(18)19)15(17)11-3-2-7-16-9-11/h2-3,7,9,12,15H,4-6,8,10H2,1H3,(H,18,19)/t12-,15-/m1/s1
Formula Weight 292.1423
Chemical Name or Material rac-trans 3'-Hydroxymethylnicotine Hemisuccinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.