missing translation for 'onlineSavingsMsg'
Learn More

rac trans-2-Phenylcyclopropylamine Hydrochloride, TRC

Product Code. 30426771
Change view
Click to view available options
Quantity:
10 mg
1 g
Unit Size:
10mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426771 1 g 1g
30410301 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426771 Supplier Toronto Research Chemicals Supplier No. TRCP3196801G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans 2-Phenylcyclopropylamine Hydrochloride
CAS 1986-47-6
Molecular Formula C9 H11 N . Cl H
Quantity 1 g
Synonym Cyclopropanamine','2-phenyl-','hydrochloride (1:1)','(1R,2S)-rel- (ACI), Cyclopropanamine','2-phenyl-','hydrochloride','(1R,2S)-rel- (9CI), Cyclopropanamine','2-phenyl-','hydrochloride','trans-(±)- (ZCI), Cyclopropylamine','2-phenyl-','hydrochloride','trans-(±)- (8CI), (±)-trans-2-Phenylcyclopropylamine hydrochloride, DL-trans-2-Phenylcyclopropylamine hydrochloride, Cyclopropanamine','2-phenyl-','hydrochloride','trans-, SKF 385 hydrochloride, trans-2-Phenylcyclopropanamine hydrochloride, trans-2-Phenylcyclopropylamine hydrochloride, Tranylcypromine hydrochloride
InChI Formula InChI=1S/C9H11N.ClH/c10-9-6-8(9)7-4-2-1-3-5-7;/h1-5,8-9H,6,10H2;1H/t8-,9+;/m1./s1
SMILES Cl.N[C@H]1C[C@@H]1c2ccccc2
IUPAC Name (1S,2R)-2-phenylcyclopropan-1-amine;hydrochloride
Molecular Weight (g/mol) 169.65
Formula Weight 169.0658
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.