missing translation for 'onlineSavingsMsg'
Learn More

rac-trans-2-Dimethylaminocyclohexanol Hydrochloride, Mikromol™

Product Code. 30052068
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30052068 25 mg 25mg
30050927 100 mg 100mg
2 options
This item is not returnable. View return policy

Product Code. 30052068

Brand: Mikromol MM0501.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

rac-trans-2-Dimethylaminocyclohexanol Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-trans-2-Dimethylaminocyclohexanol Hydrochloride
CAS 40571-56-0
Analyte or Component Names rac-trans-2-Dimethylaminocyclohexanol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C8 H17 N O . Cl H
Certifications/Compliance ISO 17025
Synonym rac-trans-2-Dimethylaminocyclohexanol Hydrochloride
InChI Formula InChI=1S/C8H17NO.ClH/c1-9(2)7-5-3-4-6-8(7)10;/h7-8,10H,3-6H2,1-2H3;1H/t7-,8-;/m1./s1
SMILES Cl.CN(C)[C@@H]1CCCC[C@H]1O
IUPAC Name (1R,2R)-2-(dimethylamino)cyclohexan-1-ol;hydrochloride
Molecular Weight (g/mol) 179.69
Quantity 25 mg
Formula Weight 179.1077 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.