missing translation for 'onlineSavingsMsg'
Learn More

rac trans-1,2-Dihydroxy-1,2-dihydronaphthalene, TRC

Product Code. 30382273
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382273 5 mg
30377404 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382273 Supplier Toronto Research Chemicals Supplier No. TRCD4527355MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-16-4
Molecular Formula C10 H10 O2
Molecular Weight (g/mol) 162.19
Synonym 1,2-Naphthalenediol, 1,2-dihydro-, (1R,2R)-rel-, 1,2-Naphthalenediol, 1,2-dihydro-, trans- (8CI), rel-(1R,2R)-1,2-Dihydro-1,2-naphthalenediol, trans-1,2-Dihydro-1,2-naphthalenediol, (±)-trans-1,2-Dihydroxy-1,2-dihydronaphthalene, 1β,2α-Dihydroxy-1,2-dihydronaphthalene, trans-1,2-Dihydroxy-1,2-dihydronaphthalene, trans-1,2-Naphthalenedihydrodiol
IUPAC Name (1S,2S)-1,2-dihydronaphthalene-1,2-diol
SMILES O[C@H]1C=Cc2ccccc2[C@@H]1O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H10O2/c11-9-6-5-7-3-1-2-4-8(7)10(9)12/h1-6,9-12H/t9-,10-/m0/s1
Formula Weight 162.0681
Chemical Name or Material (+/-)-trans-1,2-Dihydroxy-1,2-dihydronaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.