missing translation for 'onlineSavingsMsg'
Learn More

rac-threo-Dihydro Bupropion-d9 Hydrochloride, TRC

Product Code. 30382651
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382651 10 mg 10mg
30376159 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30382651 Supplier Toronto Research Chemicals Supplier No. TRCD44871710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-threo-Dihydro Bupropion-d9 Hydrochloride
CAS 1392209-60-7
Molecular Formula C13H11D9ClNO • HCl
Quantity 10 mg
Synonym 3-Chloro-α-[1-[(1,1-dimethylethyl-d9)amino]ethyl]benzenemethanol Hydrochloride, rac threo-Hydroxybupropion-d9 Hydrochloride
InChI Formula InChI=1S/C13H20ClNO.ClH/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10;/h5-9,12,15-16H,1-4H3;1H/t9-,12+;/m0./s1/i2D3,3D3,4D3;
SMILES O[C@@H](C1=CC(Cl)=CC=C1)[C@H](C)NC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H].Cl
IUPAC Name (1S,2S)-1-(3-chlorophenyl)-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]propan-1-ol;hydrochloride
Molecular Weight (g/mol) 250.813646
Formula Weight 286.1565
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.