missing translation for 'onlineSavingsMsg'
Learn More

rac threo-9,10-Dihydroxystearic Acid, TRC

Product Code. 30382039
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382039 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382039 Supplier Toronto Research Chemicals Supplier No. TRCD45475050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac threo-9,10-Dihydroxystearic Acid
CAS 2391-05-1
Molecular Formula C18 H36 O4
Quantity 50 mg
Synonym Octadecanoic acid, 9,10-dihydroxy-, (9R,10R)-rel-, Octadecanoic acid, 9,10-dihydroxy-, (R*,R*)-, Octadecanoic acid, 9,10-dihydroxy-, threo- (8CI), rel-(9R,10R)-9,10-Dihydroxyoctadecanoic acid, threo-9,10-Dihydroxyoctadecanoic acid, threo-9,10-Dihydroxystearic acid, trans-9,10-Dihydroxystearic acid
InChI Formula InChI=1S/C18H36O4/c1-2-3-4-5-7-10-13-16(19)17(20)14-11-8-6-9-12-15-18(21)22/h16-17,19-20H,2-15H2,1H3,(H,21,22)/t16-,17-/m0/s1
SMILES CCCCCCCC[C@H](O)[C@@H](O)CCCCCCCC(=O)O
IUPAC Name (9S,10S)-9,10-dihydroxyoctadecanoic acid
Molecular Weight (g/mol) 316.48
Formula Weight 316.2614
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.