missing translation for 'onlineSavingsMsg'
Learn More

rac-Terpenylic Acid, TRC

Product Code. 17992985
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17992985 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17992985 Supplier Toronto Research Chemicals Supplier No. TRCT11065025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 116-51-8
Molecular Formula C8 H12 O4
Quantity 25 mg
Synonym rac-Terpenylic Acid;
InChI Formula InChI=1S/C8H12O4/c1-8(2)5(3-6(9)10)4-7(11)12-8/h5H,3-4H2,1-2H3,(H,9,10)
SMILES CC1(C)OC(=O)CC1CC(=O)O
IUPAC Name 2-(2,2-dimethyl-5-oxooxolan-3-yl)acetic acid
Molecular Weight (g/mol) 172.179
Formula Weight 172.074
Chemical Name or Material Terpenylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.