missing translation for 'onlineSavingsMsg'
Learn More

rac Sulpiride, TRC

Product Code. 30135849
Change view
Click to view available options
Quantity:
100 mg
5 g
10 g
Unit Size:
100mg
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135849 10 g 10g
17914245 5 g 5g
17931415 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135849 Supplier Toronto Research Chemicals Supplier No. TRCS68914510G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15676-16-1
Molecular Formula C15 H23 N3 O4 S
Molecular Weight (g/mol) 341.43
Synonym Sulpiride, Benzamide, 5-(aminosulfonyl)-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxy-, o-Anisamide, N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-sulfamoyl- (8CI), (±)-Sulpiride, Abilit, Aiglonyl, Coolspan, DL-Sulpiride, Dobren, Dogmatil, Dogmatyl, Dolmatil, Eglonyl, Guastil, Meresa, Miradol, Mirbanil, Misulvan, N-(1-Ethyl-2-pyrrolidinylmethyl)-2-methoxy-5-sulfamidobenzamide, N-(1-Ethylpyrrolidin-2-ylmethyl)-2-methoxy-5-sulfamoylbenzamide, N-[(1-Ethyl-2-pyrrolidinyl)methyl]-2-methoxy-5-sulfamoylbenzamide, N-[(1-Ethyl-2-pyrrolidinyl)methyl]-5-sulfamoyl-o-anisamide, Neogama, Omperan, Pyrikappl, RD 1403, Sernevin, Splotin, Sulpirid, Sulpitil, Sulpyrid, Sursumid, Synedil, Trilan, dl-Sulpiride
IUPAC Name N-[(1-ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-sulfamoylbenzamide
SMILES CCN1CCCC1CNC(=O)c2cc(ccc2OC)S(=O)(=O)N

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H23N3O4S/c1-3-18-8-4-5-11(18)10-17-15(19)13-9-12(23(16,20)21)6-7-14(13)22-2/h6-7,9,11H,3-5,8,10H2,1-2H3,(H,17,19)(H2,16,20,21)
Formula Weight 341.1409
Chemical Name or Material Sulpiride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.