missing translation for 'onlineSavingsMsg'
Learn More

rac Styrene Oxide, TRC

Product Code. 30410846
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410846 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410846 Supplier Toronto Research Chemicals Supplier No. TRCS687795100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac Styrene Oxide
CAS 96-09-3
Molecular Formula C8H8O
Quantity 100 g
Synonym 2-Phenyloxirane, (Epoxyethyl)benzene, 1,2-Epoxy-1-phenylethane, Phenyloxirane, (Epoxyethyl)benzene, (±)-Phenylethylene Oxide, (±)-Phenyloxirane, (±)-Styrene Oxide, 1,2-Epoxyethylbenzene, 1-Phenyl-1,2-epoxyethane, 2-Phenyloxirane, Epoxystyrene, NSC 637, NSC 7582, Phenethylene Oxide, Phenylethylene Oxide, Phenyloxirane, STO, Styrene 1',2'-Oxide, Styrene 7,8-Oxide, Styrene epoxide, Styryl Oxide, alpha,beta-Epoxystyrene
InChI Formula InChI=1S/C8H8O/c1-2-4-7(5-3-1)8-6-9-8/h1-5,8H,6H2
SMILES C1OC1c2ccccc2
IUPAC Name 2-phenyloxirane
Molecular Weight (g/mol) 120.15
Formula Weight 120.0575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.