missing translation for 'onlineSavingsMsg'
Learn More

(Rac)-PT2399, MedChemExpress

Product Code. 16495166
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495166 100 mg 100mg
16436947 5 mg 5mg
16424228 10 mg 10mg
16420567 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495166 Supplier Medchem Express Supplier No. HY108697A
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress (Rac)-PT2399 (Compound 10e), the racemate of PT2399, acts as a potent and specific hypoxia-inducible factor 2a (HIF-2α) inhibitor with an IC50 of 0.01 μM.

(Rac)-PT2399 (Compound 10e), the racemate of PT2399, acts as a potent and specific hypoxia-inducible factor 2a (HIF-2α) inhibitor with an IC50 of 0.01 μM.

Specifications

Chemical Name or Material (Rac)-PT2399
CAS 1672662-07-5
Color White
Purity Grade Notes Research
Molecular Formula C17H10F5NO4S
Quantity 100 mg
Solubility Information DMSO : 120 mg/mL (286.18 mM; Need ultrasonic)
SMILES N#CC1=CC(F)=CC(OC2=CC=C(S(=O)(C(F)F)=O)C3=C2CC(F)(F)C3O)=C1
Molecular Weight (g/mol) 419.32
Formula Weight 419.32
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.