missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac-Norsertraline.HCl, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
rac-Norsertraline.HCl
Specifications
Specifications
| Chemical Name or Material | Norsertraline hydrochloride |
| CAS | 91797-57-8 |
| Analyte or Component Names | Norsertraline hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H15 Cl2 N . Cl H |
| Synonym | (±)-cis-N-Desmethylsertraline Hydrochloride, cis-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride, (±)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride, rac-Norsertraline Hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.N[C@H]1 CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13 |
| IUPAC Name | (1 S,4 S)-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction