missing translation for 'onlineSavingsMsg'
Learn More

rac Normetanephrine Hydrochloride, TRC

Product Code. 18011430
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18011430 50 mg 50mg
17929739 25 mg 25mg
18043830 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18011430 Supplier Toronto Research Chemicals Supplier No. TRCN72150050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1011-74-1
Molecular Formula C9 H13 N O3 . Cl H
Molecular Weight (g/mol) 219.67
Synonym Benzenemethanol, α-(aminomethyl)-4-hydroxy-3-methoxy-, hydrochloride (1:1), Benzenemethanol, α-(aminomethyl)-4-hydroxy-3-methoxy-, hydrochloride (9CI), Benzenemethanol, α-(aminomethyl)-4-hydroxy-3-methoxy-, hydrochloride, (±)-, Vanillyl alcohol, α-(aminomethyl)-, hydrochloride, (±)- (8CI), (±)-Normetanephrine hydrochloride, (±)-α-(Aminomethyl)-4-hydroxy-3-methoxybenzyl alcohol hydrochloride, DL-Normetanephrine hydrochloride, Normetadrenaline hydrochloride, Normetanephrine hydrochloride, dl-Normetanephrine hydrochloride, 3-O-Methylnoradrenaline Hydrochloride
IUPAC Name 4-(2-amino-1-hydroxyethyl)-2-methoxyphenol;hydrochloride
SMILES Cl.COc1cc(ccc1O)C(O)CN

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H13NO3.ClH/c1-13-9-4-6(8(12)5-10)2-3-7(9)11;/h2-4,8,11-12H,5,10H2,1H3;1H
Formula Weight 219.0662
Percent Purity >95%
Chemical Name or Material Normetanephrine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.