missing translation for 'onlineSavingsMsg'
Learn More

rac-Norcamphor, Mikromol™

Product Code. 30049862
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049862 25 mg 25mg
17931296 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049862 Supplier Mikromol Supplier No. MM0103.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

rac-Norcamphor
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Norcamphor
CAS 497-38-1
Analyte or Component Names Norcamphor
Shipping Condition Room Temperature
Molecular Formula C7 H10 O
Certifications/Compliance ISO 9001
Synonym 2-Norbornanone (6CI,8CI), 2,5-Methanocyclohexanone, 2-Oxonorbornane, NSC 66537, NSC 92359, Norcamphor, Racemic norcamphor, dl-Norcamphor, Bicyclo[2.2.1]heptan-2-one, (+/-)-2-Norbornanone, (+/-)-Norcamphor
InChI Formula InChI=1S/C7H10O/c8-7-4-5-1-2-6(7)3-5/h5-6H,1-4H2
SMILES O=C1CC2CCC1C2
IUPAC Name bicyclo[2.2.1]heptan-3-one
Molecular Weight (g/mol) 110.15
Quantity 25 mg
Formula Weight 110.0732 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.