missing translation for 'onlineSavingsMsg'
Learn More

rac N,O-Didesmethyl Tramadol-d3, TRC

Product Code. 30381059
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381059 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30381059 Supplier Toronto Research Chemicals Supplier No. TRCD4412221MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1261398-22-4
Molecular Formula C14H18D3NO2
Quantity 1 mg
Synonym N,O-Didesmethyltramadol-D3 (N-methyl-D3), N,O-Didesmethyltramadol-d3, rel-3-[(1R,2R)-1-Hydroxy-2-[[(methyl-d3)amino]methyl]cyclohexyl]phenol, rac N,O-Didesmethyl Tramadol-d3, 3-[(1RS,2RS)-1-Hydroxy-2-[[(methyl-d3)amino]methyl]cyclohexyl]phenol, cis-3-[1-Hydroxy-2-[[(methyl-d3)amino]methyl]cyclohexyl]phenol
InChI Formula InChI=1S/C14H21NO2/c1-15-10-12-5-2-3-8-14(12,17)11-6-4-7-13(16)9-11/h4,6-7,9,12,15-17H,2-3,5,8,10H2,1H3/t12-,14+/m1/s1/i1D3
SMILES [2H]C([2H])([2H])NC[C@H]1CCCC[C@]1(O)c2cccc(O)c2
Molecular Weight (g/mol) 238.34
Formula Weight 238.1761
Chemical Name or Material (+/-)-N,O-Di-Desmethyl Tramadol-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.